Learn More
Chemglass Life Sciences Plug, Drosophila, High Density Foam, for Wide Vials
SDP
Catalog No. 501897637
Encompass
• Fly plugs are made of high-density foam• Causes minimal shedding and reduce evaporation• Maintains tight fit after repeated use and removal• Plugs are compatible with CGE-4789 Drosophila vials and bottles (Sold Separately)
Catalog No. 50-189-7637 Supplier Chemglass Life Sciences Supplier No. CGE4790003
• Fly plugs are made of high-density foam• Causes minimal shedding and reduce evaporation• Maintains tight fit after repeated use and removal• Plugs are compatible with CGE-4789 Drosophila vials and bottles (Sold Separately)