Learn More
Med Vet International Ear Plugs 3M E-A-R Classic Cordless One Size Fits Most Yellow, 310-1001, 2000 Pairs
Catalog No. 502830136
Encompass_Preferred
Slow-recovery foam helps ensure comfort and minimize pressure Bright yellow color provides a quick visual compliance check Dermatologically safe foam is non-irritating, nonflammable, and moisture-resistant Convenient pillow pak keeps each pair clean
Catalog No. 50-283-0136 Supplier Med Vet International Supplier No. 650987CS
Slow-recovery foam helps ensure comfort and minimize pressure Bright yellow color provides a quick visual compliance check Dermatologically safe foam is non-irritating, nonflammable, and moisture-resistant Convenient pillow pak keeps each pair clean