Learn More
Med Vet International Ear Plugs 3M E-A-Rsoft Yellow Neons Cordless One Size Fits Most Yellow, 312-1250, 200 Pairs
Catalog No. 502829407
Encompass_Preferred
Regular size fits most ear canals Soft foam earplugs are easy to roll down and insert, then expand within the ear canal for maximum comfort Noise reduction rating (NRR)* 33 dB. CSA Class ALPoly bag keeps each pair of earplugs clean before use
Catalog No. 50-282-9407 Supplier Med Vet International Supplier No. 765622BX
Regular size fits most ear canals Soft foam earplugs are easy to roll down and insert, then expand within the ear canal for maximum comfort Noise reduction rating (NRR)* 33 dB. CSA Class ALPoly bag keeps each pair of earplugs clean before use