Learn More
Med Vet International Ear Plugs 3M E-A-Rsoft Yellow Neons Cordless One Size Fits Most Yellow, 312-1250, 2000 Pairs
Catalog No. 502831479
Encompass_Preferred
Regular size fits most ear canals Soft foam earplugs are easy to roll down and insert, then expand within the ear canal for maximum comfort Noise reduction rating (NRR)* 33 dB. CSA Class ALPoly bag keeps each pair of earplugs clean before use
Catalog No. 50-283-1479 Supplier Med Vet International Supplier No. 765622CS
Regular size fits most ear canals Soft foam earplugs are easy to roll down and insert, then expand within the ear canal for maximum comfort Noise reduction rating (NRR)* 33 dB. CSA Class ALPoly bag keeps each pair of earplugs clean before use