Learn More
Ace Glass, Inc. PLUG FOR -42 PLUG FOR -42
SDP
Supplier:  Ace Glass, Inc. 781844
Encompass_Preferred
Plug for Mercury Reservoir, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-979-478