Learn More
Microsolv Technology Corporation Plug for Mobile Phase Cap, replacement. PEEK 1/4-28" threads. 1 EA.
Catalog No. 502271522
Encompass_Preferred
Knurled for a tight grip and also has "flat" surfaces for using a wrench to quarter tighten for best seal or to loosen if plug gets stuck. Screw thread for 1/4 - 28" holes.
This item is not returnable. View return policy
Catalog No. 50-227-1522 Supplier Microsolv Technology Corporation Supplier No. 4902501N
Knurled for a tight grip and also has "flat" surfaces for using a wrench to quarter tighten for best seal or to loosen if plug gets stuck. Screw thread for 1/4 - 28" holes.