Learn More
MSC Balax Plug Gage, No-Go, 5/8-11, Class 3B, 1 ea.
Catalog No. 5000511327
Encompass
This 5/8-11 thread plug gage is a precision tool designed for inspecting internal threads. Constructed from high-speed steel with a bright surface finish, this No-Go gage is used to verify that the pitch diameter of a threaded hole does not exceed the maximum allowable size for a Class 3B fit.
  • Class 3B fit for high-precision thread inspection
  • Constructed from high-speed steel for durability
  • Bright surface finish
  • No-Go configuration for accurate size verification
  • Thread size of 5/8-11
  • Thread length of 0.5 in.
  • Overall length of 38.1 mm (1.5 in.)
Catalog No. 50-005-11327 Supplier MSC Supplier No. 05386131
This 5/8-11 thread plug gage is a precision tool designed for inspecting internal threads. Constructed from high-speed steel with a bright surface finish, this No-Go gage is used to verify that the pitch diameter of a threaded hole does not exceed the maximum allowable size for a Class 3B fit.
  • Class 3B fit for high-precision thread inspection
  • Constructed from high-speed steel for durability
  • Bright surface finish
  • No-Go configuration for accurate size verification
  • Thread size of 5/8-11
  • Thread length of 0.5 in.
  • Overall length of 38.1 mm (1.5 in.)