Learn More
MSC Balax Thread Plug Gage, 3/4-10 Thread, 3B Class, No Go, 1 ea
Catalog No. 5000510338
Encompass
This single-end plug thread gage is designed for precise inspection of internal thread accuracy. Constructed from durable high-speed tool steel with a bright finish, this gage features a 3/4-10 thread size and 3B class tolerance. It is specifically engineered for No Go applications to ensure adherence to strict thread specifications.
  • Manufactured from high-speed tool steel
  • Hardness rating of 63-65 Rockwell
  • Single-end configuration with No Go designation
  • Number 3 taperlock handle size
  • Overall length of 1-1/2 in. with 1/2 in. thread length
  • Made in the USA
Catalog No. 50-005-10338 Supplier MSC Supplier No. 05387121
This single-end plug thread gage is designed for precise inspection of internal thread accuracy. Constructed from durable high-speed tool steel with a bright finish, this gage features a 3/4-10 thread size and 3B class tolerance. It is specifically engineered for No Go applications to ensure adherence to strict thread specifications.
  • Manufactured from high-speed tool steel
  • Hardness rating of 63-65 Rockwell
  • Single-end configuration with No Go designation
  • Number 3 taperlock handle size
  • Overall length of 1-1/2 in. with 1/2 in. thread length
  • Made in the USA