Learn More
MSC Balax Thread Plug Gage, 5/8-18 Thread, 2B and 3B Class, Go, 1 ea
Catalog No. 5000516931
Encompass
This thread plug gage is designed for precision inspection of internal threads, ensuring components meet specified tolerances. Constructed from high speed tool steel with a bright finish, this gage provides durability and consistent measurement accuracy in metalworking applications.
  • Designed for 5/8-18 thread size
  • Compatible with 2B and 3B classes
  • Single end configuration for go verification
  • Hardness rating of 63-65 Rockwell
  • Features a #3 taperlock handle
  • Thread length of 7/8 in. and overall length of 2-9/16 in.
Catalog No. 50-005-16931 Supplier MSC Supplier No. 05386180
This thread plug gage is designed for precision inspection of internal threads, ensuring components meet specified tolerances. Constructed from high speed tool steel with a bright finish, this gage provides durability and consistent measurement accuracy in metalworking applications.
  • Designed for 5/8-18 thread size
  • Compatible with 2B and 3B classes
  • Single end configuration for go verification
  • Hardness rating of 63-65 Rockwell
  • Features a #3 taperlock handle
  • Thread length of 7/8 in. and overall length of 2-9/16 in.