Learn More
MSC Balax Thread Plug Gage, 3/4-16 Thread, 2B and 3B Class, High Speed Tool Steel, 1 ea
Catalog No. 5000516924
Encompass
This single end, go plug thread gage is designed for precise measurement of 3/4-16 thread sizes. Constructed from high speed tool steel with a bright finish, this gage provides durability and wear resistance in demanding inspection environments.
  • Designed for 3/4-16 thread size and 2B and 3B class of fit
  • Hardness rating of 63-65 Rockwell
  • Single end, go configuration
  • Features a #3 taperlock handle
  • Thread length of 7/8 in.
  • Overall length of 2-3/4 in.
  • Made in the USA
Catalog No. 50-005-16924 Supplier MSC Supplier No. 05387162
This single end, go plug thread gage is designed for precise measurement of 3/4-16 thread sizes. Constructed from high speed tool steel with a bright finish, this gage provides durability and wear resistance in demanding inspection environments.
  • Designed for 3/4-16 thread size and 2B and 3B class of fit
  • Hardness rating of 63-65 Rockwell
  • Single end, go configuration
  • Features a #3 taperlock handle
  • Thread length of 7/8 in.
  • Overall length of 2-3/4 in.
  • Made in the USA