Learn More
MSC Balax Thread Plug Gage, 3/8-24, 2B and 3B Class, Go, 1 ea
Catalog No. 5000516858
Encompass
This single-end Go thread plug gage is designed for precision inspection of internal threads. Constructed from high speed tool steel with a bright finish, it offers durability and accuracy for industrial quality control applications.
  • Designed for 3/8-24 thread size
  • Compatible with 2B and 3B classes
  • Features 63-65 Rockwell hardness
  • Includes #2 taperlock handle size
  • Constructed from high speed tool steel
  • Total overall length of 2 in. with 3/4 in. thread length
  • Made in the USA
Catalog No. 50-005-16858 Supplier MSC Supplier No. 05382247
This single-end Go thread plug gage is designed for precision inspection of internal threads. Constructed from high speed tool steel with a bright finish, it offers durability and accuracy for industrial quality control applications.
  • Designed for 3/8-24 thread size
  • Compatible with 2B and 3B classes
  • Features 63-65 Rockwell hardness
  • Includes #2 taperlock handle size
  • Constructed from high speed tool steel
  • Total overall length of 2 in. with 3/4 in. thread length
  • Made in the USA