Learn More
MSC Balax Thread Plug Gage, 1 in.-8 Thread, 2B Class, No Go, 1 ea.
Catalog No. 5000516859
Encompass
This No Go thread plug gage is designed for precision inspection of internal threads, ensuring they meet the specified 2B class of fit requirements. Constructed from high speed tool steel and hardened to 63-65 Rockwell, this gage provides durability and consistent measurement accuracy for demanding metalworking applications.
  • Manufactured from high speed tool steel for increased durability
  • Hardened to 63-65 Rockwell to maintain precision over time
  • Features a single end design with a bright finish
  • Equipped with a #4 taperlock handle
  • Designed for use as a no go gage for 1 in.-8 threads
Catalog No. 50-005-16859 Supplier MSC Supplier No. 05389093
This No Go thread plug gage is designed for precision inspection of internal threads, ensuring they meet the specified 2B class of fit requirements. Constructed from high speed tool steel and hardened to 63-65 Rockwell, this gage provides durability and consistent measurement accuracy for demanding metalworking applications.
  • Manufactured from high speed tool steel for increased durability
  • Hardened to 63-65 Rockwell to maintain precision over time
  • Features a single end design with a bright finish
  • Equipped with a #4 taperlock handle
  • Designed for use as a no go gage for 1 in.-8 threads