Learn More
MSC Balax Thread Plug Gage, 5/16-18, 3B Class, No Go, 1 ea.
Catalog No. 5000512393
Encompass
This No Go thread plug gage is designed for precision inspection of internal threads, ensuring adherence to specified tolerance standards. Constructed from high speed tool steel with a bright finish, this gage provides durability and consistent performance in demanding measurement applications.
  • Thread size: 5/16-18
  • Class of fit: 3B
  • Hardness: 63-65 Rockwell
  • Handle size: #1 Taperlock
  • Thread length: 5/16 in.
  • Overall length: 1-5/16 in.
  • Neck length: 9/16 in.
  • Material: High speed tool steel
Catalog No. 50-005-12393 Supplier MSC Supplier No. 05381207
This No Go thread plug gage is designed for precision inspection of internal threads, ensuring adherence to specified tolerance standards. Constructed from high speed tool steel with a bright finish, this gage provides durability and consistent performance in demanding measurement applications.
  • Thread size: 5/16-18
  • Class of fit: 3B
  • Hardness: 63-65 Rockwell
  • Handle size: #1 Taperlock
  • Thread length: 5/16 in.
  • Overall length: 1-5/16 in.
  • Neck length: 9/16 in.
  • Material: High speed tool steel