Learn More
Grainger Plug In Elbow 1/4 in PK10
Catalog No. 502763775
Encompass_Preferred
The Parker Plug-In Elbow Composite is a 90 degree elbow fitting made of composite material. It has 1/4 inch NPT female pipe thread connections and is rated for a maximum pressure of 500 psi at 70°F. The elbow has an operating temperature range of -40°F to 180°F. It measures 2-1/2 inches in length and weighs 0.19 lbs. The elbow fitting provides corrosion resistance and has a glossy black finish. It is designed for use in pneumatic conveying systems to provide a leak-tight 90 degree turn. This elbow fitting can also be used with air, water, oil and inert gas applications. It is manufactured in the USA by Parker Hannifin Corporation.
Catalog No. 50-276-3775 Supplier Grainger Supplier No. 16W616
The Parker Plug-In Elbow Composite is a 90 degree elbow fitting made of composite material. It has 1/4 inch NPT female pipe thread connections and is rated for a maximum pressure of 500 psi at 70°F. The elbow has an operating temperature range of -40°F to 180°F. It measures 2-1/2 inches in length and weighs 0.19 lbs. The elbow fitting provides corrosion resistance and has a glossy black finish. It is designed for use in pneumatic conveying systems to provide a leak-tight 90 degree turn. This elbow fitting can also be used with air, water, oil and inert gas applications. It is manufactured in the USA by Parker Hannifin Corporation.