Learn More
Grainger Plug In Elbow 1/4 In PK5
Catalog No. 502764311
Encompass_Preferred
The Parker 16W617 Plug-In Elbow Composite is a 90 degree elbow fitting made of composite material. It has 1/4 inch NPT female pipe thread connections and is rated for a maximum pressure of 3000 psi at 70°F. This elbow fitting is lightweight and corrosion resistant. It measures 2.19 inches long x 1.5 inches wide and weighs 0.1 lbs. The 16W617 elbow fitting from Parker is designed for use in hydraulic and pneumatic systems in industrial applications.
Catalog No. 50-276-4311 Supplier Grainger Supplier No. 16W617
The Parker 16W617 Plug-In Elbow Composite is a 90 degree elbow fitting made of composite material. It has 1/4 inch NPT female pipe thread connections and is rated for a maximum pressure of 3000 psi at 70°F. This elbow fitting is lightweight and corrosion resistant. It measures 2.19 inches long x 1.5 inches wide and weighs 0.1 lbs. The 16W617 elbow fitting from Parker is designed for use in hydraulic and pneumatic systems in industrial applications.