Learn More
Grainger TORK Plug-In Timer, 1 min Min Time
Catalog No. NC2132299
Encompass_Preferred
The TORK Plug-In Timer is an electronic timer that can be plugged directly into an outlet to control power to a connected device. It has a digital LED display and buttons to program the on/off times. The timer has a minimum on/off time of 1 minute and a maximum of 12 hours. It operates on 120V AC and has a 15A resistive rating. The timer measures 2.4 x 4.5 x 1.4 inches. This plug-in timer provides automated control of power to lamps, fans, and other electronic devices on a schedule.
This item is not returnable. View return policy
Catalog No. NC2132299 Supplier Grainger Supplier No. 60NP78
The TORK Plug-In Timer is an electronic timer that can be plugged directly into an outlet to control power to a connected device. It has a digital LED display and buttons to program the on/off times. The timer has a minimum on/off time of 1 minute and a maximum of 12 hours. It operates on 120V AC and has a 15A resistive rating. The timer measures 2.4 x 4.5 x 1.4 inches. This plug-in timer provides automated control of power to lamps, fans, and other electronic devices on a schedule.