Learn More
Chemglass Life Sciences Chemglass Plug Only, 0-8 mm PTFE-Tipped Replacement Plug for PTFE-Tip Valves
SDP
Catalog No. 502733513
Encompass_Preferred
Replacement PTFE-tipped plug designed for use with PTFE-tip valve assemblies. This "plug only" spare fits valve port diameters from 0-8 mm and provides chemical resistance and a sealed closure for compatible valve bodies.
  • Ptfe tip for broad chemical resistance.
  • Designed for port diameters 0-8 mm.
  • Intended as a replacement plug for compatible PTFE-tip valve assemblies.
  • Sold individually for spare and maintenance needs.
Catalog No. 50-273-3513 Supplier Chemglass Life Sciences Supplier No. CGK56102
Replacement PTFE-tipped plug designed for use with PTFE-tip valve assemblies. This "plug only" spare fits valve port diameters from 0-8 mm and provides chemical resistance and a sealed closure for compatible valve bodies.
  • Ptfe tip for broad chemical resistance.
  • Designed for port diameters 0-8 mm.
  • Intended as a replacement plug for compatible PTFE-tip valve assemblies.
  • Sold individually for spare and maintenance needs.