Learn More
Chemglass Life Sciences 2mm 120° PTFE Plug Only, 15.2/30
SDP
Catalog No. 501531735
Encompass_Preferred
2mm 120¿ PTFE Plug Only, 15.2/30. Modified Dean-Stark receiver designed for solvents that are either lighter or heavier than water. The lower three-way PTFE stopcock allows for the withdraw or return of the distillate during the distillation process. With a 10/30 outer joint on the top of the vapor tube for a 3¿ immersion thermometer. Graduations are the same as CG-1261.
Catalog No. 50-153-1735 Supplier Chemglass Life Sciences Supplier No. CG44064
2mm 120¿ PTFE Plug Only, 15.2/30. Modified Dean-Stark receiver designed for solvents that are either lighter or heavier than water. The lower three-way PTFE stopcock allows for the withdraw or return of the distillate during the distillation process. With a 10/30 outer joint on the top of the vapor tube for a 3¿ immersion thermometer. Graduations are the same as CG-1261.