Learn More
Chemglass Life Sciences 2mm Glass Plug Only, 12/30
SDP
Supplier:  Chemglass Life Sciences CG41091
Encompass_Preferred
2mm Glass Plug Only, 12/30. Stopcock has three sidearms sealed 90 apart. The solid glass plug has a "T" shaped bore so that any two or all three sidearms can be connected at one time. All sizes are supplied complete with a metal retaining device CG-423.
Catalog No. 50-153-1731