Learn More
Chemglass Life Sciences Stopcock, High Vacuum T-Bore, 2mm Glass Plug ONLY, 12/30 Plug Size
SDP
Catalog No. 501531759
Encompass_Preferred
Stopcock, High Vacuum T-Bore, 2mm Glass Plug ONLY, 12/30 Plug Size. Similar to CG-462, but with three stems sealed 90 apart. Plug only.
Catalog No. 50-153-1759 Supplier Chemglass Life Sciences Supplier No. CG46401PO
Stopcock, High Vacuum T-Bore, 2mm Glass Plug ONLY, 12/30 Plug Size. Similar to CG-462, but with three stems sealed 90 apart. Plug only.