Learn More
Waters Corp Nylon Column Plug, for Storage, 1 per Pack
Catalog No. NC3672519 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These column plugs are crafted from durable nylon, specifically designed to provide secure storage for complete columns. They help to protect and preserve the integrity of your columns when not in active use.
  • Made from nylon material.
  • Designed for secure column storage.
  • Supplied individually.
Catalog No. NC3672519 Supplier Waters Corp Supplier No. WAT015674
These column plugs are crafted from durable nylon, specifically designed to provide secure storage for complete columns. They help to protect and preserve the integrity of your columns when not in active use.
  • Made from nylon material.
  • Designed for secure column storage.
  • Supplied individually.