Learn More
Microsolv Technology Corporation Mat, Sealing, Chemical Resistant Polymer, Pre-Slit, Plug Style for 96, 350ul Inserts. 1 EA.
Catalog No. 502270561
Encompass_Preferred
Made of a chemically resistant polymer, this Mat can be used with typical HPLC organic solvents. Designed and molded to seal up to 96 each 350ul Inserts in the U-2D Micro-Sample Management System. This mat is Pre-Slit and has alphanumeric markings on the top side of the mats, along the edges for identification of the Inserts.
This item is not returnable. View return policy
Catalog No. 50-227-0561 Supplier Microsolv Technology Corporation Supplier No. 9529630REA
Made of a chemically resistant polymer, this Mat can be used with typical HPLC organic solvents. Designed and molded to seal up to 96 each 350ul Inserts in the U-2D Micro-Sample Management System. This mat is Pre-Slit and has alphanumeric markings on the top side of the mats, along the edges for identification of the Inserts.