Learn More
Microsolv Technology Corporation Mats, Sealing, Large, Chemical Resistant Polymer, Pre-Slit, Plug Style for 96, 500 or 1000ul Inserts. 5/CS.
Catalog No. 502270567
Encompass_Preferred
Mats are made of a chemically resistant polymer that can be used with typical HPLC organic solvents. Designed and molded to seal up to 96 each 500ul or 1ml Inserts in the U-2D Micro-Sample Management System. These mats are Pre-Slit and have alphanumeric markings on the top side of the mats, along the edges for identification of the Inserts.
This item is not returnable. View return policy
Catalog No. 50-227-0567 Supplier Microsolv Technology Corporation Supplier No. 95296LG30RCS
Mats are made of a chemically resistant polymer that can be used with typical HPLC organic solvents. Designed and molded to seal up to 96 each 500ul or 1ml Inserts in the U-2D Micro-Sample Management System. These mats are Pre-Slit and have alphanumeric markings on the top side of the mats, along the edges for identification of the Inserts.