Learn More
Microsolv Technology Corporation Mats, Sealing, Large, Silicone Rubber, Pre-Slit, Plug Style for 96, 500 and 1000ul Inserts. 5/CS.
Catalog No. 502270009
Encompass_Preferred
Mats are made of ultra pure Silicone Rubber, designed and molded to seal up to 96 each 500ul and 1ml Inserts in the U-2D Micro-Sample Management System. These mats are Pre-Slit and have alphanumeric markings on the top side of the mats, along the edges for identification of the Inserts. Use with Aqueous Solutions.
This item is not returnable. View return policy
Catalog No. 50-227-0009 Supplier Microsolv Technology Corporation Supplier No. 95296LG30PCS
Mats are made of ultra pure Silicone Rubber, designed and molded to seal up to 96 each 500ul and 1ml Inserts in the U-2D Micro-Sample Management System. These mats are Pre-Slit and have alphanumeric markings on the top side of the mats, along the edges for identification of the Inserts. Use with Aqueous Solutions.