Learn More
Microsolv Technology Corporation Mat, Sealing, Silicone Rubber, Pre-Slit, Plug Style for 96, 350ul Inserts. 1 EA.
Catalog No. 502271097
Encompass_Preferred
Made of ultra pure Silicone Rubber, this mat is designed and molded to seal up to 96 each 350ul Inserts in the U-2D Micro-Sample Management System. This mat is Pre-Slit and has alphanumeric markings on the top side of the mat, along the edges for identification of the Inserts.
This item is not returnable. View return policy
Catalog No. 50-227-1097 Supplier Microsolv Technology Corporation Supplier No. 9529630PEA
Made of ultra pure Silicone Rubber, this mat is designed and molded to seal up to 96 each 350ul Inserts in the U-2D Micro-Sample Management System. This mat is Pre-Slit and has alphanumeric markings on the top side of the mat, along the edges for identification of the Inserts.