Learn More
Instech Stainless steel catheter / tubing plug, 20ga x 12mm. Nonsterile. Package of 100.
SDP
Catalog No. 5081224
Encompass
The Instech SP20/12 is a stainless steel catheter plug used to seal externalized catheters prior to infusion or sampling procedures. Sized at 20ga and 12mm in length, the plug provides a secure, low‑dead‑volume closure that protects catheter patency and prevents fluid loss. The rounded edges help minimize tubing damage, and the plugs are supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 20ga stainless steel catheter plug
  • 12mm length for secure catheter sealing
  • Rounded edges to protect tubing and catheter tips
  • Non‑sterile; packaged in sets of 100
  • Used to temporarily close externalized catheters before infusion
Catalog No. 50-812-24 Supplier Instech Supplier No. SP20/12
The Instech SP20/12 is a stainless steel catheter plug used to seal externalized catheters prior to infusion or sampling procedures. Sized at 20ga and 12mm in length, the plug provides a secure, low‑dead‑volume closure that protects catheter patency and prevents fluid loss. The rounded edges help minimize tubing damage, and the plugs are supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 20ga stainless steel catheter plug
  • 12mm length for secure catheter sealing
  • Rounded edges to protect tubing and catheter tips
  • Non‑sterile; packaged in sets of 100
  • Used to temporarily close externalized catheters before infusion