Learn More
Instech Stainless steel catheter / tubing plug, 22ga x 12mm. Nonsterile. Package of 100.
SDP
Catalog No. 5081226
Encompass
The Instech SP22/12 is a stainless steel catheter plug used to seal externalized catheters prior to infusion or sampling procedures. Sized at 22ga and 12mm in length, it provides a secure, low‑dead‑volume closure that protects catheter patency and prevents fluid leakage. Its rounded edges help reduce tubing wear, and it is supplied non‑sterile in packs of 100 for laboratory infusion system assembly.
Features
  • 22ga stainless steel catheter plug
  • 12mm length for secure catheter sealing
  • Rounded edges to minimize tubing damage
  • Non‑sterile; packaged in sets of 100
  • Used to close externalized catheters before infusion
Catalog No. 50-812-26 Supplier Instech Supplier No. SP22/12
The Instech SP22/12 is a stainless steel catheter plug used to seal externalized catheters prior to infusion or sampling procedures. Sized at 22ga and 12mm in length, it provides a secure, low‑dead‑volume closure that protects catheter patency and prevents fluid leakage. Its rounded edges help reduce tubing wear, and it is supplied non‑sterile in packs of 100 for laboratory infusion system assembly.
Features
  • 22ga stainless steel catheter plug
  • 12mm length for secure catheter sealing
  • Rounded edges to minimize tubing damage
  • Non‑sterile; packaged in sets of 100
  • Used to close externalized catheters before infusion