Learn More
Instech Stainless steel catheter / tubing plug, 23ga x 12mm. Nonsterile. Package of 100.
SDP
Catalog No. 5081227
Encompass
The Instech SP23/12 is a stainless steel catheter plug designed to seal externalized catheters before infusion or sampling procedures. Sized at 23ga and 12mm in length, it provides a secure, low‑dead‑volume closure to maintain catheter patency and prevent fluid leakage. Rounded edges help protect delicate tubing, and the plugs are supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 23ga stainless steel catheter plug
  • 12mm length for secure catheter sealing
  • Rounded edges to minimize tubing damage
  • Non‑sterile; packaged in sets of 100
  • Used to close externalized catheters prior to infusion
Catalog No. 50-812-27 Supplier Instech Supplier No. SP23/12
The Instech SP23/12 is a stainless steel catheter plug designed to seal externalized catheters before infusion or sampling procedures. Sized at 23ga and 12mm in length, it provides a secure, low‑dead‑volume closure to maintain catheter patency and prevent fluid leakage. Rounded edges help protect delicate tubing, and the plugs are supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 23ga stainless steel catheter plug
  • 12mm length for secure catheter sealing
  • Rounded edges to minimize tubing damage
  • Non‑sterile; packaged in sets of 100
  • Used to close externalized catheters prior to infusion