Learn More
Instech Stainless steel catheter / tubing plug, 25ga x 12mm. Nonsterile. Package of 100.
SDP
Catalog No. 5081228
Encompass
The Instech SP25/12 is a stainless steel catheter plug used to seal externalized catheters prior to infusion or sampling procedures. Sized at 25ga and 12mm in length, it provides a secure, low‑dead‑volume closure that helps maintain catheter patency and prevent fluid leakage. Rounded edges protect delicate tubing, and the plugs are supplied non‑sterile in packs of 100 for use in laboratory infusion systems.
Features
  • 25ga stainless steel catheter plug
  • 12mm length for secure catheter sealing
  • Rounded edges reduce risk of tubing damage
  • Non‑sterile; supplied in packs of 100
  • Used to close externalized catheters before infusion
Catalog No. 50-812-28 Supplier Instech Supplier No. SP25/12
The Instech SP25/12 is a stainless steel catheter plug used to seal externalized catheters prior to infusion or sampling procedures. Sized at 25ga and 12mm in length, it provides a secure, low‑dead‑volume closure that helps maintain catheter patency and prevent fluid leakage. Rounded edges protect delicate tubing, and the plugs are supplied non‑sterile in packs of 100 for use in laboratory infusion systems.
Features
  • 25ga stainless steel catheter plug
  • 12mm length for secure catheter sealing
  • Rounded edges reduce risk of tubing damage
  • Non‑sterile; supplied in packs of 100
  • Used to close externalized catheters before infusion