Learn More
IDEX Health & Science Upchurch PLUG TEFZEL FOR M-6 THREADS
Catalog No. NC1919059
Encompass_Preferred
Idex Threaded Plugs and Caps, Black, Tefzel (ETFE), Port Type: M6 Flat Bottom.
Catalog No. NC1919059 Supplier IDEX Health & Science Upchurch Supplier No. P314
Idex Threaded Plugs and Caps, Black, Tefzel (ETFE), Port Type: M6 Flat Bottom.