Learn More
McMaster-Carr 1/4" Pipe Size Plugs
Catalog No. NC1457083
Encompass
The McMaster-Carr 1/4" NPT Pipe Plug is made of 18-8 stainless steel for corrosion resistance. It has a hex drive for tightening and is rated for pressures up to 10,000 psi. The plug has a knurled end for easy finger tightening and loosening. It meets ASME B1.20.1 standards and is RoHS compliant. The plug is made in USA and sold individually.
Catalog No. NC1457083 Supplier McMaster-Carr Supplier No. 1901K59
The McMaster-Carr 1/4" NPT Pipe Plug is made of 18-8 stainless steel for corrosion resistance. It has a hex drive for tightening and is rated for pressures up to 10,000 psi. The plug has a knurled end for easy finger tightening and loosening. It meets ASME B1.20.1 standards and is RoHS compliant. The plug is made in USA and sold individually.