Learn More
Trajan Scientific VICI Valco Plug, Cheminert, PEEK, 1/4-28, can be used to Close Off an unused Port in a 1/4-28 Valve or Manifold, 5 Pack
Catalog No. 501090313
Encompass
VICI Valco Plug, Cheminert, PEEK, 1/4-28, can be used to Close Off an unused Port in a 1/4-28 Valve or Manifold, 5 Pack. Made in the USA.
Catalog No. 50-109-0313 Supplier Trajan Scientific Supplier No. CPPK
VICI Valco Plug, Cheminert, PEEK, 1/4-28, can be used to Close Off an unused Port in a 1/4-28 Valve or Manifold, 5 Pack. Made in the USA.