Learn More
Cole-Parmer Cole-Parmer PTFE Thread tape (Plumber's), 1/2"w x 43ft
Supplier:  Cole-Parmer 0827033
Encompass_Preferred
PTFE Plumber's Tape is used on all types of threaded fittings for passage of potable and non-potable water, most chemicals, oxygen, gas, hydraulic and high-pressure lines. Withstands harsh temperatures from -450 to 550F (-267 to 287C). Strong chemical resistance. Stretches 50% without breaking.
Catalog No. NC9033595