Learn More
Specifications
Specifications
| Color Indicator | White/Yellow to Pink |
| Format | Sheet |
| For Use With (Application) | For detection of Lead, lead ions |
| Quantity | 40 Pack |
| Test Time | 2 min. |
| Type | Qualitative Test Paper |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
