Learn More
Aviva Systems Biology Corp PLUNC Antibody - middle region (ARP42475_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42475_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLUNC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PLUNC. Alternate names: LUNX, NASG, SPLUNC1, SPURT, bA49G10.5, PLUNC, LPLUNC3ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53336