Learn More
Waters Corp Waters Plunger, 0.0787 in. Diameter, 1.415 in. Length, Sapphire
Catalog No. 502039441 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement plunger assembly is designed for use in various solvent managers and UPLC systems. It is constructed from sapphire for durability and precise operation.
  • Has a diameter of 0.0787 in.
  • Features a length of 1.415 in.
  • Constructed from sapphire material.
  • Supplied as a package of two units.
  • Recommended for annual replacement.
  • Utilized on solvent managers.
  • Compatible with a wide range of UPLC systems.
Catalog No. 50-203-9441 Supplier Waters Corp Supplier No. 700002600
This replacement plunger assembly is designed for use in various solvent managers and UPLC systems. It is constructed from sapphire for durability and precise operation.
  • Has a diameter of 0.0787 in.
  • Features a length of 1.415 in.
  • Constructed from sapphire material.
  • Supplied as a package of two units.
  • Recommended for annual replacement.
  • Utilized on solvent managers.
  • Compatible with a wide range of UPLC systems.