Learn More
Optimize Technologies Plunger, Waters 510, 590, 600/600E, 610, 6K/6KA
SDP
Catalog No. 50114175
Encompass_Preferred
Pistons are, by design, a consumable part of the HPLC pump and should be replaced on a regular preventative maintenance schedule. To maintain peak performance and avoid unplanned downtime, sapphire pistons require inspection every six months with yearly or as-needed replacement. Optimize sapphire pistons offer optimal concentricity and resistance to wear. Each sapphire rod is specifically cut in relation to the grain of the crystal lattice, yielding maximal resistance to breakage and superior wear characteristics. Attachment of the rod to the piston ferrule is performed through a proprietary process ensuring concentricity. This is an important, often overlooked procedure parameter that has a direct effect on seal lifetime. This specific piston is designed to work with Waters 510, 590, 600/600E. 610, and 6K/6KA pump models. It is equivalent to OEM part number WAT025656.
Catalog No. 50-114-175 Supplier Optimize Technologies Supplier No. 393401188
Pistons are, by design, a consumable part of the HPLC pump and should be replaced on a regular preventative maintenance schedule. To maintain peak performance and avoid unplanned downtime, sapphire pistons require inspection every six months with yearly or as-needed replacement. Optimize sapphire pistons offer optimal concentricity and resistance to wear. Each sapphire rod is specifically cut in relation to the grain of the crystal lattice, yielding maximal resistance to breakage and superior wear characteristics. Attachment of the rod to the piston ferrule is performed through a proprietary process ensuring concentricity. This is an important, often overlooked procedure parameter that has a direct effect on seal lifetime. This specific piston is designed to work with Waters 510, 590, 600/600E. 610, and 6K/6KA pump models. It is equivalent to OEM part number WAT025656.