Learn More
Waters Corp Plunger Assembly, .125 Dia, UPC2, Sapphire
Catalog No. 502391972 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement Plunger Assembly used in UPC2 Binary Solvent Managers.
  • Units per package: 2 pk
  • Type: Plunger
  • System type: UPC2
  • Material: Sapphire
  • Replacement schedule: As required
Catalog No. 50-239-1972 Supplier Waters Corp Supplier No. 700009000
This is a replacement Plunger Assembly used in UPC2 Binary Solvent Managers.
  • Units per package: 2 pk
  • Type: Plunger
  • System type: UPC2
  • Material: Sapphire
  • Replacement schedule: As required