Learn More
Waters Corp Waters Plunger Assembly, For 1525μ HPLC Pump, With Brass Seat
Catalog No. 50793548 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The plunger assembly is a critical replacement component designed for use with 1525μ HPLC pumps. It plays a vital role in maintaining optimal pump performance and is recommended for regular replacement during routine maintenance or when specific operational issues arise.
  • Designed for 1525μ HPLC pump systems.
  • Includes a brass seat for proper functionality.
  • Recommended for replacement semi-annually or when experiencing erratic retention times.
  • Helps resolve issues such as solvent leaks from the pump head.
  • Addresses large pressure fluctuations to ensure stable operation.
  • Requires careful handling due to its delicate nature.
  • Can be cleaned by sonication in a 50:50 methanol/water solution.
Catalog No. 50-793-548 Supplier Waters Corp Supplier No. WAT030547
The plunger assembly is a critical replacement component designed for use with 1525μ HPLC pumps. It plays a vital role in maintaining optimal pump performance and is recommended for regular replacement during routine maintenance or when specific operational issues arise.
  • Designed for 1525μ HPLC pump systems.
  • Includes a brass seat for proper functionality.
  • Recommended for replacement semi-annually or when experiencing erratic retention times.
  • Helps resolve issues such as solvent leaks from the pump head.
  • Addresses large pressure fluctuations to ensure stable operation.
  • Requires careful handling due to its delicate nature.
  • Can be cleaned by sonication in a 50:50 methanol/water solution.