Learn More
Waters Corp Waters Plunger Assembly, Sapphire, 0.280 in. Diameter
Catalog No. 50792597 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Plunger Assembly is a replacement part constructed from sapphire. It moves with the piston, filling the pump head with solvent through the inlet check valve when it retracts, and expelling the solvent from the pump head when it extends forward. It has a diameter of 0.280 in. and is used on 2525/2535/2545 Pumps.
  • Constructed from sapphire
  • Features a diameter of 0.280 in.
  • Compatible with 2525/2535/2545 Pumps
  • Recommended to change seals when changing plungers
Catalog No. 50-792-597 Supplier Waters Corp Supplier No. 700001532
The Plunger Assembly is a replacement part constructed from sapphire. It moves with the piston, filling the pump head with solvent through the inlet check valve when it retracts, and expelling the solvent from the pump head when it extends forward. It has a diameter of 0.280 in. and is used on 2525/2535/2545 Pumps.
  • Constructed from sapphire
  • Features a diameter of 0.280 in.
  • Compatible with 2525/2535/2545 Pumps
  • Recommended to change seals when changing plungers