Learn More
Waters Corp Waters Corporation Plunger Assembly, HPLC
Catalog No. 50791791 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This plunger assembly is a crucial component in High-Performance Liquid Chromatography (HPLC) systems, designed to enhance system reliability and extend the life of critical parts. It works by actively lubricating the plunger and removing contaminants that could compromise seal integrity.
  • Optimizes system uptime.
  • Extends the lifespan of plunger seals.
  • Extends the lifespan of plungers.
  • Lubricates the plunger.
  • Flushes away solvents or precipitated salts from the piston chamber.
  • Beneficial for applications using buffered mobile phases.
Catalog No. 50-791-791 Supplier Waters Corp Supplier No. 410002205
This plunger assembly is a crucial component in High-Performance Liquid Chromatography (HPLC) systems, designed to enhance system reliability and extend the life of critical parts. It works by actively lubricating the plunger and removing contaminants that could compromise seal integrity.
  • Optimizes system uptime.
  • Extends the lifespan of plunger seals.
  • Extends the lifespan of plungers.
  • Lubricates the plunger.
  • Flushes away solvents or precipitated salts from the piston chamber.
  • Beneficial for applications using buffered mobile phases.