Learn More
Pipette Supplies, Inc. PLUNGER BUTTON NEW STYLE 20UL
SDP
Catalog No. NC1568162
Encompass
Newer Style Plunger Button with ridged edge for:Gilson Pipetman Classic 20μL (P20)Made in France
Catalog No. NC1568162 Supplier Pipette Supplies, Inc. Supplier No. GI F144783
Newer Style Plunger Button with ridged edge for:Gilson Pipetman Classic 20μL (P20)Made in France