Learn More
Trajan Scientific Plunger Diamond MS Medical 1/EA
SDP
Supplier:  Trajan Scientific 0355193
Encompass_Preferred
Replacement plunger for 100 L CTC RTC and Thermo RSH Diamond MS syringe
Catalog No. 50-004-94112