Learn More
Trajan Scientific Plunger Diamond MS Medical 1/EA
SDP
Catalog No. 5000494720
Encompass_Preferred
Replacement plunger for 10 L CTC RTC and Thermo RSH Diamond MS syringe
Catalog No. 50-004-94720 Supplier Trajan Scientific Supplier No. 0355190
Replacement plunger for 10 L CTC RTC and Thermo RSH Diamond MS syringe