Learn More
Trajan Scientific Plunger Gas Tight GT Headspace 1/EA
SDP
Supplier:  Trajan Scientific 032846
Encompass_Preferred
Replacement P2 5MF-RTC/RSH headspace energized gas tight 2 5 mL plunger PK1
Catalog No. 50-004-94018