Learn More
Trajan Scientific Plunger Gas Tight GT Medical 2/PK
SDP
Catalog No. 5000494806
Encompass_Preferred
P10R-C/T gas tight 10 L plunger PK2
Catalog No. 50-004-94806 Supplier Trajan Scientific Supplier No. 0318121
P10R-C/T gas tight 10 L plunger PK2