Learn More
Trajan Scientific Plunger Medical Life Science Lab 1/EA
SDP
Catalog No. 5000494866
Encompass_Preferred
Replacement plunger for 10 L removable needle Varian 8035 8100 and 8200 syringe with gas tight plunger and 5 3 cm 0 50 mm OD side hole needle
Catalog No. 50-004-94866 Supplier Trajan Scientific Supplier No. 031218
Replacement plunger for 10 L removable needle Varian 8035 8100 and 8200 syringe with gas tight plunger and 5 3 cm 0 50 mm OD side hole needle