Learn More
Neta Scientific Agilent Plunger seal for 1100/1200 and 1050 2/PK
SDP
Catalog No. 50924552
Encompass_Preferred
This item is manufactured by Agilent
Agilent Piston seal, PTFE, used with G1311A/B/C, G1312A/B/C, G1310A/B For G1310A, G1311A, G1312A, G1376A, G2226A, 1120, 1220, G1310B, G1311B, G1312B, G1311C, G1312C, G4302A - Instrument Parts
UNSPSC Code  41105106
- AA
Catalog No. 50-924552 Supplier Neta Scientific Supplier No. 50636589
This item is manufactured by Agilent
Agilent Piston seal, PTFE, used with G1311A/B/C, G1312A/B/C, G1310A/B For G1310A, G1311A, G1312A, G1376A, G2226A, 1120, 1220, G1310B, G1311B, G1312B, G1311C, G1312C, G4302A - Instrument Parts
UNSPSC Code  41105106
- AA