Learn More
Waters Corp Plunger Seal Insertion Tool (100 µL)
Supplier:  Waters Corp WAT024765
Encompass_Preferred
The Plunger Seal Insertion Tool is used to pry the old plunger seal out of the pump head, as well as aligning the new plunger seal correctly into the pump head.
Catalog No. 50-793-514