Learn More
Waters Corp Waters Plunger Seal Insertion Tool
Supplier:  Waters Corp WAT270969
Encompass_Preferred
The Plunger Seal Insertion Tool is designed to facilitate the easy and precise insertion of new seals. This tool is used once the new seals have been lubricated with 100% alcohol, ensuring proper installation.
  • Provides an easy way to insert new seals.
  • Ensures precise insertion of new seals.
  • Suitable for use after seals are lubricated with 100% alcohol.
Catalog No. 50-793-824